Learn More
Erlab ChemTrap H402 Filtration with BE Carbon filtration Option for Acid, or Corrosive Safety Cabinets Midcap Version
Shop All Erlab Products
Click to view available options
Includes:
BE Carbon Filter
Description
- Erlab™'s ChemTrap Filtration Option for Safety Cabinets is a filtration system that will filter the interior space of acids, or corrosive cabinets.
- The filters capture and retain toxic fumes, then return clean air back to the laboratory, eliminating odors when the door is opened.
- Designed as a standalone system with no need for ducting to the outside.
- User protection while saving energy
- Easy installation with connection via bung port
- No ductwork needed
- Eliminates toxic fumes and odors while purifying lab air
- Adaptable to any cabinet
- Greener Choice Claims
- Environmental benefits include:
- Free-of a substance that causes environmental harm
- Non-Toxic
- Ozone-Safe or Safe for the Ozone Layer
- Renewable Materials
Specifications
Specifications
Includes | BE Carbon Filter |
Product Type | Cabinet Shelf |