Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHERP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541813
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA541813 100 μL
1 options

Catalog No. PIPA541813

Supplier: Invitrogen™ PA541813

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SERLLAAVEA FYSPPSHDRP RNSEGWEQNG LYEFFRAKMR ARRRKGQEKR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.

CHERP is involved in calcium homeostasis, growth and proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHERP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CHERP
Gene Accession No. Q8IWX8
Gene Alias 5730408I11Rik; Calcium homeostasis endoplasmic reticulum protein; CHERP; D8Wsu96e; DAN16; DAN26; ERPROT 213-21; ERPROT213-21; fc83d01; ik:tdsubc_2h12; protein with polyglutamine repeat; Scaf6; spliceosome associated factor; SCAF6; SRA1; SR-related CTD associated factor 6; SR-related CTD-associated factor 6; tdsubc_2h12; Unknown (protein for IMAGE:8005617); wu:fc83d01; xx:tdsubc_2h12; zgc:55518
Gene Symbols CHERP
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CHERP.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10523
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.