Learn More
Invitrogen™ CHERP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597238
Description
Immunogen sequence: LPAGLMAPLV KLEDHEYKPL DPKDIRLPPP MPPSERLLAA VEAFYSPPSH DRPRNSEGWE QNGLYEFFRA KMRARRRKGQ EKRNSGPSRS RSRSKSRGRS S; Positive Samples: HeLa, HepG2, A-549, Jurkat, Mouse testis; Cellular Location: Cytoplasm, Endoplasmic reticulum, perinuclear region.
CHERP is involved in calcium homeostasis, growth and proliferation.
Specifications
CHERP | |
Polyclonal | |
Unconjugated | |
CHERP | |
5730408I11Rik; Calcium homeostasis endoplasmic reticulum protein; CHERP; D8Wsu96e; DAN16; DAN26; ERPROT 213-21; ERPROT213-21; fc83d01; ik:tdsubc_2h12; protein with polyglutamine repeat; Scaf6; spliceosome associated factor; SCAF6; SRA1; SR-related CTD associated factor 6; SR-related CTD-associated factor 6; tdsubc_2h12; Unknown (protein for IMAGE:8005617); wu:fc83d01; xx:tdsubc_2h12; zgc:55518 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10523, 27967, 290614 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CGZ0, Q8IWX8 | |
CHERP | |
A synthetic peptide corresponding to a sequence within amino acids 650-750 of human CHERP (NP_0063783). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.