Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHERP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103674 Supplier Invitrogen™ Supplier No. PA5103674
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CHERP.

CHERP is involved in calcium homeostasis, growth and proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHERP
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHERP
Gene Accession No. Q8CGZ0, Q8IWX8
Gene Alias 5730408I11Rik; Calcium homeostasis endoplasmic reticulum protein; CHERP; D8Wsu96e; DAN16; DAN26; ERPROT 213-21; ERPROT213-21; fc83d01; ik:tdsubc_2h12; protein with polyglutamine repeat; Scaf6; spliceosome associated factor; SCAF6; SRA1; SR-related CTD associated factor 6; SR-related CTD-associated factor 6; tdsubc_2h12; Unknown (protein for IMAGE:8005617); wu:fc83d01; xx:tdsubc_2h12; zgc:55518
Gene Symbols CHERP
Host Species Rabbit
Immunogen A synthesized peptide derived from human CHERP(Accession Q8IWX8), corresponding to amino acid residues D423-N473.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10523, 27967
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.