Learn More
CHERP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304620AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
CHERP is involved in calcium homeostasis, growth and proliferation.Specifications
CHERP | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8IWX8 | |
CHERP | |
Between 400 and 450 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CHERP | |
DAN16, ERPROT 213-21, ERPROT213-21, protein with polyglutamine repeat, SCAF6, SRA1, SR-related CTD associated factor 6, SR-related CTD-associated factor 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10523 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.