Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHERP Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501568213
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8213 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8213 Supplier Bethyl Laboratories, Inc Supplier No. A304621AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

CHERP is involved in calcium homeostasis, growth and proliferation.

Specifications

Antigen CHERP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CHERP
Gene Accession No. Q8IWX8
Gene Alias DAN16, ERPROT 213-21, ERPROT213-21, protein with polyglutamine repeat, SCAF6, SRA1, SR-related CTD associated factor 6, SR-related CTD-associated factor 6
Gene Symbols CHERP
Host Species Rabbit
Immunogen Between 866 and 916
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10523
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.