Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHERP Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CHERP |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4884
![]() ![]() |
Proteintech Group Inc
253891AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
CHERP gene ontology annotations related to this gene include negative regulation of cell proliferation; positive regulation of calcineurin-NFAT signaling cascade; release of sequestered calcium ion into cytosol; RNA processing.Specifications
CHERP | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8CGZ0, Q8IWX8 | |
10523, 27967 | |
CHERP Fusion Protein Ag13428 | |
Primary | |
-20°C | |
CHERP |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CHERP, DAN16, DAN26, ERPROT 213 21, SCAF6, SRA1 | |
Cherp | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title