Learn More
Eisco™ Chert Mineral Specimen
SCI_ED
Catalog No. S27596 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27596 12/Pk.
S27574 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27596 Supplier Eisco™ Supplier No. ESNG0067PK12
Add to Cart
Add to Cart
Sometimes referred to as flint, Chert is a sedimentary rock rich in silica.
  • Correlates well to Next Generation Science Standards Evidence
  • Chert is perfect for teaching about the many forms of quartz, hardness, crystal habit, and fracture
Specifications
Product Type Mineral
Grade K to 16
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.1 kg