Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHES1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204051JF646
Description
CHES1 Polyclonal antibody specifically detects CHES1 in Human samples. It is validated for Western BlotSpecifications
CHES1 | |
Polyclonal | |
Western Blot | |
C14orf116, Checkpoint suppressor 1PRO1635, CHES1forkhead box protein N3, chromosome 14 open reading frame 116, forkhead box N3 | |
The immunogen recognized by this antibody maps to a region between residue 440 and 490 of human Forkhead Box N3 using the numbering given in entry NP_001078940.1 (GeneID 1112). | |
0.1 mL | |
Cell Cycle and Replication | |
1112 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction