Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHES1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP238866
Description
CHES1 Polyclonal specifically detects CHES1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CHES1 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
O00409 | |
FOXN3 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MGPVMPPSKKPESSGISVSSGLSQCYGGSGFSKALQEDDDLDFSLPDIRLEEGAMEDEELT | |
0.1 mL | |
Cell Cycle and Replication | |
1112 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C14orf116, Checkpoint suppressor 1PRO1635, CHES1forkhead box protein N3, chromosome 14 open reading frame 116, forkhead box N3 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction