Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHFR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542327
Description
Peptide sequence: REWTIGRRRG CDLSFPSNKL VSGDHCRIVV DEKSGQVTLE DTSTSGTVIN Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
E3 ubiquitin-protein ligase that functions in the antephase checkpoint by actively delaying passage into mitosis in response to microtubule poisons. Acts in early prophase before chromosome condensation, when the centrosome move apart from each other along the periphery of the nucleus. Probably involved in signaling the presence of mitotic stress caused by microtubule poisons by mediating the 'Lys-48'-linked ubiquitination of target proteins, leading to their degradation by the proteasome. Promotes the ubiquitination and subsequent degradation of AURKA and PLK1. Probably acts as a tumor suppressor, possibly by mediating the polyubiquitination of HDAC1, leading to its degradation. May also promote the formation of 'Lys-63'-linked polyubiquitin chains and functions with the specific ubiquitin-conjugating UBC13-MMS2 (UBE2N-UBE2V2) heterodimer. Substrates that are polyubiquitinated at 'Lys-63' are usually not targeted for degradation, but are rather involved in signaling cellular stress.
Specifications
CHFR | |
Polyclonal | |
Unconjugated | |
Chfr | |
5730484M20Rik; C230082M18; checkpoint with forkhead and ring finger domains; Checkpoint with forkhead and RING finger domains protein; checkpoint with forkhead and ring finger domains, E3 ubiquitin protein ligase; Chfr; E3 ubiquitin-protein ligase CHFR; fc43f10; FLJ10796; FLJ33629; RING finger protein 196; RING-type E3 ubiquitin transferase CHFR; RNF116; RNF196; si:dkey-69h6.7; wu:fc43f10 | |
Rabbit | |
Affinity chromatography | |
RUO | |
55743 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96EP1 | |
Chfr | |
Synthetic peptide directed towards the N-terminal of human CHFR (aa 36-85). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction