Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHFR Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550177

Catalog No. PA550177


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total CHFR protein.

E3 ubiquitin-protein ligase that functions in the antephase checkpoint by actively delaying passage into mitosis in response to microtubule poisons. Acts in early prophase before chromosome condensation, when the centrosome move apart from each other along the periphery of the nucleus. Probably involved in signaling the presence of mitotic stress caused by microtubule poisons by mediating the 'Lys-48'-linked ubiquitination of target proteins, leading to their degradation by the proteasome. Promotes the ubiquitination and subsequent degradation of AURKA and PLK1. Probably acts as a tumor suppressor, possibly by mediating the polyubiquitination of HDAC1, leading to its degradation. May also promote the formation of 'Lys-63'-linked polyubiquitin chains and functions with the specific ubiquitin-conjugating UBC13-MMS2 (UBE2N-UBE2V2) heterodimer. Substrates that are polyubiquitinated at 'Lys-63' are usually not targeted for degradation, but are rather involved in signaling cellular stress.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CHFR
Polyclonal
Unconjugated
Chfr
5730484M20Rik; C230082M18; checkpoint with forkhead and ring finger domains; Checkpoint with forkhead and RING finger domains protein; checkpoint with forkhead and ring finger domains, E3 ubiquitin protein ligase; Chfr; E3 ubiquitin-protein ligase CHFR; fc43f10; FLJ10796; FLJ33629; RING finger protein 196; RING-type E3 ubiquitin transferase CHFR; RNF116; RNF196; si:dkey-69h6.7; wu:fc43f10
Rabbit
Antigen affinity chromatography
RUO
231600, 288734, 55743
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q810L3, Q96EP1
Chfr
Synthesized peptide derived from Internal of human CHFR.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.