Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHFR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587905
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587905 Supplier Invitrogen™ Supplier No. PA587905
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MERPEEGKQS PPPQPWGRLL RLGAEEGEPH VLLRKREWTI GRRRGCDLSF PSNKLVSGDH CRIVVDEKSG QVTLEDTSTS GTVINKLKVV KKQTCPLQTG DVIYLVYRKN EPEHNVAYLY ESLSEKQGMT QESFDTSGAG AGRGADPRVP PSSPATQVCF EEPQPSTSTS DLFPTASASS TEPSPAGRER SSSCGSGGGG ISPKGSGPSV ASDEVSSFAS ALPDRKTASF SSLEPQDQED LEPVKKKMRG DGDLDLNGQL LVAQPRRNAQ TVHEDVRAAA GKPDKMEETL; Positive Samples: HT-1080, 22Rv1, HepG2, Mouse kidney, Mouse testis; Cellular Location: Nucleus, PML body.

E3 ubiquitin-protein ligase that functions in the antephase checkpoint by actively delaying passage into mitosis in response to microtubule poisons. Acts in early prophase before chromosome condensation, when the centrosome move apart from each other along the periphery of the nucleus. Probably involved in signaling the presence of mitotic stress caused by microtubule poisons by mediating the 'Lys-48'-linked ubiquitination of target proteins, leading to their degradation by the proteasome. Promotes the ubiquitination and subsequent degradation of AURKA and PLK1. Probably acts as a tumor suppressor, possibly by mediating the polyubiquitination of HDAC1, leading to its degradation. May also promote the formation of 'Lys-63'-linked polyubiquitin chains and functions with the specific ubiquitin-conjugating UBC13-MMS2 (UBE2N-UBE2V2) heterodimer. Substrates that are polyubiquitinated at 'Lys-63' are usually not targeted for degradation, but are rather involved in signaling cellular stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHFR
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Chfr
Gene Accession No. Q810L3, Q96EP1
Gene Alias 5730484M20Rik; C230082M18; checkpoint with forkhead and ring finger domains; Checkpoint with forkhead and RING finger domains protein; checkpoint with forkhead and ring finger domains, E3 ubiquitin protein ligase; Chfr; E3 ubiquitin-protein ligase CHFR; fc43f10; FLJ10796; FLJ33629; RING finger protein 196; RING-type E3 ubiquitin transferase CHFR; RNF116; RNF196; si:dkey-69h6.7; wu:fc43f10
Gene Symbols Chfr
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human CHFR (NP_0011548181).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231600, 55743
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.