Learn More
CHGA Mouse anti-Human, Mouse, Clone: OTI2E12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF506098

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Chromogranin A (CHGA) is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. It is found in secretory vesicles of neurons and endocine cells. CHGA is a precursor to three biologically active peptides: vasostatin, pancreastatin, and parastatin. These peptides act as autocine or paracrine negative modulators of the neuroendocrine system. CHGA is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.Specifications
CHGA | |
Monoclonal | |
Unconjugated | |
CHGA | |
CGA | |
Mouse | |
Affinity Chromatography | |
RUO | |
1113, 12652 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI2E12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P10645, P26339 | |
CHGA | |
Full length human recombit protein of human CHGA produced in HEK293T cell. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.