Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHI3L1 Monoclonal Antibody (A3G13)
GREENER_CHOICE

Catalog No. PIMA544972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544972 Supplier Invitrogen™ Supplier No. MA544972
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CHI3L1 Monoclonal Antibody for Western Blot

Human cartilage glycoprotein 39 (GP-39), also known as YKL-40, is a glycoprotein secreted by articular chondrocytes, synoviocytes and macrophages. Serum and synovial fluid GP-39 levels are elevated in inflammatory diseases and correlate with the degree of joint destruction in rheumatoid arthritis. GP-39 is expressed in articular chondrocytes and synovial cells, as well as in liver, but is undetectable in muscle tissues, lung, pancreas, mononuclear cells and fibroblasts. GP-39 is a candidate autoantigen in rheumatoid arthritis and is important in the capacity of cells to respond to and cope with changes in their environment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHI3L1
Applications Western Blot
Classification Monoclonal
Clone A3G13
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene CHI3L1
Gene Accession No. P36222
Gene Alias 38 kDa heparin-binding glycoprotein; 38kDa heparin-binding glycoprotein; 39 kDa synovial protein; 39 kDa whey protein; 40 kDa secretory signalling glycoprotein; ASRT7; AW208766; BP40; BP40 precursor; breast regression protein 39; Brp39; BRP39 protein; Cartilage glycoprotein 39; cartilage glycoprotein-39; CGP39; CGP-39; CHI3L1; Chil1; chitinase 3 like 1; Chitinase 3 like protein 1CGP 39; chitinase 3-like 1; chitinase 3-like 1 (cartilage glycoprotein-39); chitinase 3-like 1 protein; chitinase-3-like protein 1; chitinase-like 1; chitinase-like protein 1; CLP-1; DKFZp686N19119; FLJ38139; GP38K; Gp39; GP-39; hCGP39; HC-gp39; hCGP-39; HCGP-3P; Mammary gland protein 40; MGP-40; signal processing protein; signal-processing protein; SPC-40; SPS-40; YKL40; YKL-40; YYL-40
Gene Symbols CHI3L1
Host Species Mouse
Immunogen Recombinant protein within extracellular domainof human CHI3L1.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1116
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.