Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHI3L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 120361AP150UL

Description
Human cartilage glycoprotein 39 (GP-39), also known as YKL-40, is a glycoprotein secreted by articular chondrocytes, synoviocytes and macrophages. Serum and synovial fluid GP-39 levels are elevated in inflammatory diseases and correlate with the degree of joint destruction in rheumatoid arthritis. GP-39 is expressed in articular chondrocytes and synovial cells, as well as in liver, but is undetectable in muscle tissues, lung, pancreas, mononuclear cells and fibroblasts. GP-39 is a candidate autoantigen in rheumatoid arthritis and is important in the capacity of cells to respond to and cope with changes in their environment.Specifications
CHI3L1 | |
Polyclonal | |
Unconjugated | |
CHI3L1 | |
39 kDa synovial protein, ASRT7, Cartilage glycoprotein 39, CGP 39, CHI3L1, Chitinase 3 like protein 1, GP 39, GP39, HC gp39, hCGP 39, HCGP 3P, YKL 40, YKL40, YYL 40 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1116, 12654, 89824 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P36222, Q61362, Q9WTV1 | |
Chi3l1 | |
CHI3L1 Fusion Protein Ag2710 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction