Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHIA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530526
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530526 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530526 Supplier Invitrogen™ Supplier No. PA530526
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: JurKat. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CHIA degrades chitin and chitotriose. May participate in the defense against nematodes and other pathogens. There are 3 named isoforms produced by alternative splicing. CHIA is induced via a T helper-2 (Th2)-specific, interleukin-13-mediated pathway in epithelial cells and macrophages. CHIA may be an important mediator of IL13-induced responses in Th2-dominated disorders such as asthma. CHIA hydrolysis N-acetyl-beta-D-glucosaminide 1,4-beta-linkages in chitin and chitodextrins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHIA
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CHIA
Gene Accession No. Q9BZP6
Gene Alias 2200003E03Rik; acidic chitinase; acidic mammalian chitinase; AMCASE; Chia; Chia1; CHIT2; chitinase, acidic; chitinase, acidic 1; eosinophil chemotactic cytokine; lung-specific protein TSA1902; small acid mammalian chitinase; TSA1902; YNL
Gene Symbols CHIA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CHIA. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27159
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.