Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHIC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542660 Supplier Invitrogen™ Supplier No. PA542660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LRRYAPDPVL VRGAGHITVF GLSNKFDTEF PSVLTGKVAP EEFKTSIGRV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

This gene encodes a cysteine-rich hydrophobic (CHIC) domain-containing protein, and is one of the few protein-coding genes found near the X-inactivation center. Studies in mouse indicate that the mouse ortholog of this gene is subject to X-inactivation in mouse. Experiments with other CHIC domain-containing family members show that the cysteine residues are palmitoylated post-translationally, resulting in membrane association. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHIC1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CHIC1
Gene Accession No. Q5VXU3
Gene Alias Brain X-linked protein; Brx; CHIC1; cysteine rich hydrophobic domain 1; cysteine-rich hydrophobic domain 1; cysteine-rich hydrophobic domain 1 protein; cysteine-rich hydrophobic domain-containing protein 1; cystein-rich hydrophobic domain 1; Unknown (protein for IMAGE:7449829); wu:fj33g02
Gene Symbols CHIC1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CHIC1 (aa 81-130).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53344
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.