Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHIC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110926
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110926 100 μL
PIPA5111741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5110926 Supplier Invitrogen™ Supplier No. PA5110926
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SIEKLLEWEN NRLYHKLCLH WRLSKRKCET N Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

This gene encodes a member of the CHIC family of proteins. The encoded protein contains a cysteine-rich hydrophobic (CHIC) motif, and is localized to vesicular structures and the plasma membrane. This gene is associated with some cases of acute myeloid leukemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHIC2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHIC2
Gene Accession No. Q9UKJ5
Gene Alias 1700081B18Rik; 4930502K01Rik; BrX-like translocated in leukemia; BTL; Chic2; cysteine rich hydrophobic domain 2; cysteine-rich hydrophobic domain 2; cysteine-rich hydrophobic domain 2 protein; cysteine-rich hydrophobic domain-containing protein 2; cystein-rich hydrophobic domain 2; MGC21173; zgc:55670
Gene Symbols CHIC2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CHIC2. Recombinant protein control fragment (Product #RP-90088).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26511
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.