Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANXA5, Chicken, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89129493 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-493 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-493 Supplier Abnova Corporation Supplier No. PAB27978
Add to Cart
Add to Cart

Chicken polyclonal antibody against synthetic peptide of ANXA5

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29kb containing 13 exons, and encodes a single transcript of approximately 1.6kb and a protein product with a molecular weight of about 35kDa. [provided by RefSeq

Sequence: ETSGNLEQLLLAVVK

Specifications

Antigen ANXA5
Applications Immuno-MRM (Multiple Reaction Monitoring)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Description Chicken polyclonal antibody against synthetic peptide of ANXA5
Formulation In Phosphate-Buffered Saline with 0.02% Sodium Azide.
Gene ANXA5
Gene Alias ANX5/ENX2/PP4
Gene Symbols ANXA5
Host Species Chicken
Immunogen A Synthetic peptide corresponding to amino acids of ANXA5
Purification Method Antigen affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.