Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C3, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130603 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-603 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-603 Supplier Abnova Corporation Supplier No. PAB29121
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against synthetic peptides of Bovine C3.

Sequence: KSSAYAAFQYRC
KEKNRWEEPNQKLC

Specifications

Antigen C3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against synthetic peptides of Bovine C3.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.05% sodium azide).
Gene C3
Gene Symbols C3
Host Species Chicken
Immunogen Two synthetic peptides (conjugated with KLH) correspoding to Bovine C3 emulsified in Freund's adjuvant.
Purification Method IgY fraction
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280677
Target Species Bovine
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.