Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD276, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130615 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-615 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-615 Supplier Abnova Corporation Supplier No. PAB29169

Chicken polyclonal antibody raised against native Human CD276.

Costimulatory B7 molecules (e.g., B7-1, or CD80; MIM 112203) signal through CD28 (MIM 186760) family molecules such as CD28, CTLA4 (MIM 123890), and ICOS (MIM 604558).[supplied by OMIM

Specifications

Antigen CD276
Applications Flow Cytometry
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against native Human CD276.
Dilution Flow Cytometry (5-20 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.075% sodium azide)
Gene CD276
Gene Alias B7-H3/B7H3
Gene Symbols CD276
Host Species Chicken
Immunogen A synthetic peptide corresponding to the extracellular domains of human CD276 fused to the hinge region and the CH2 and CH3 domains of human IgG1 (described in Leitner et al., EJI, 2009 39(7) 1754-64).
Purification Method IgY Fraction
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80381
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.