Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNP, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130569 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-569 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-569 Supplier Abnova Corporation Supplier No. PAB29066
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against recombinant Human CNP.

Specifications

Antigen CNP
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against recombinant Human CNP.
Dilution Immunocytochemistry(1:1000-1:2000) Immunohistochemistry(1:1000-1:2000) Western Blot(1:2000-1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% sodium azide)
Gene Accession No. NP_149124
Gene Alias CNP1
Gene Symbols CNP
Host Species Chicken
Immunogen A KLH-conjugated synthetic peptide corresponding to a region of CNP gene product shared between the human (NP_149124, NCBI) and mouse (P16330, NCBI) sequences.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1267/12799
Target Species Human, Mouse
Content And Storage Store at 4°C and avoid from light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.