Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRP, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130606 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-606 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-606 Supplier Abnova Corporation Supplier No. PAB29128

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chicken polyclonal antibody raised against native dog CRP.

Specifications

Antigen CRP
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against native dog CRP.
Dilution ELISA (1:100-1:300 for capture) Western Blot (1:1000 - 1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide).
Gene CRP
Gene Symbols CRP
Host Species Chicken
Immunogen Native purified dog CRP from dog serum with Freund's adjuvant.
Purification Method Immunoaffinity Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 488629
Target Species Dog
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.