Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRYM, Chicken, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89129475 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-475 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-475 Supplier Abnova Corporation Supplier No. PAB27960
Add to Cart
Add to Cart

Chicken polyclonal antibody against synthetic peptide of CRYM

Crystallins are separated into two classes: taxon-specific and ubiquitous. The former class is also called phylogenetically-restricted crystallins. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. This gene encodes a taxon-specific crystallin protein that binds NADPH and has sequence similarity to bacterial ornithine cyclodeaminases. The encoded protein does not perform a structural role in lens tissue, and instead it binds thyroid hormone for possible regulatory or developmental roles. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq

Sequence: EAVLYVDSQEAALK

Specifications

Antigen CRYM
Applications Immuno-MRM (Multiple Reaction Monitoring)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Description Chicken polyclonal antibody against synthetic peptide of CRYM
Formulation In Phosphate-Buffered Saline with 0.02% Sodium Azide.
Gene CRYM
Gene Alias DFNA40/THBP
Gene Symbols CRYM
Host Species Chicken
Immunogen A Synthetic peptide corresponding to amino acids of CRYM
Purification Method Antigen affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1428
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.