Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCHSD1, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89127388 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-388 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-388 Supplier Abnova Corporation Supplier No. PAB25764
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against synthetic peptide of FCHSD1.

Specifications

Antigen FCHSD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against synthetic peptide of FCHSD1.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene FCHSD1
Gene Accession No. NP_258260
Gene Alias FLJ00007/NWK2
Gene Symbols FCHSD1
Host Species Chicken
Immunogen A synthetic peptide corresponding to 15 amino acids at C-terminus of human FCHSD1.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89848
Test Specificity Multiple isoforms of FCHSD1 are known to exist. FCHSD1 antibody is predicted to not cross-react with FCHSD2.
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.