Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP10, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130617 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-617 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-617 Supplier Abnova Corporation Supplier No. PAB29171
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against native Human FKBP10.

The protein encoded by this gene belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase family. It is located in endoplasmic reticulum and acts as molecular chaperones. An alternatively spliced variant encoding different isoform has been found, but the biological validity of the variant is not determined. [provided by RefSeq

Specifications

Antigen FKBP10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against native Human FKBP10.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.05% sodium azide)
Gene FKBP10
Gene Alias FKBP6/FKBP65/FLJ20683/FLJ22041/FLJ23833/hFKBP65
Gene Symbols FKBP10
Host Species Chicken
Immunogen Native purified FKBP10 from normal human third trimester placenta by reversed-order OFarrell gel electrophoresis (ROOGE).
Purification Method IgY Fraction
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60681
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.