Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FN1, Chicken, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89129478 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-478 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-478 Supplier Abnova Corporation Supplier No. PAB27963
Add to Cart
Add to Cart

Chicken polyclonal antibody against synthetic peptide of FN1

This gene encodes fibronectin, a glycoprotein present in a soluble dimeric form in plasma, and in a dimeric or multimeric form at the cell surface and in extracellular matrix. Fibronectin is involved in cell adhesion and migration processes including embryogenesis, wound healing, blood coagulation, host defense, and metastasis. The gene has three regions subject to alternative splicing, with the potential to produce 20 different transcript variants. However, the full-length nature of some variants has not been determined. [provided by RefSeq

Sequence: IYLYTLNDNAR

Specifications

Antigen FN1
Applications Immuno-MRM (Multiple Reaction Monitoring)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Description Chicken polyclonal antibody against synthetic peptide of FN1
Formulation In Phosphate-Buffered Saline with 0.02% Sodium Azide.
Gene FN1
Gene Alias CIG/DKFZp686F10164/DKFZp686H0342/DKFZp686I1370/DKFZp686O13149/ED-B/FINC/FN/FNZ/GFND/GFND2/LETS/MSF
Gene Symbols FN1
Host Species Chicken
Immunogen A Synthetic peptide corresponding to amino acids of FN1
Purification Method Antigen affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2335
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.