Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD1, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130576 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-576 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-576 Supplier Abnova Corporation Supplier No. PAB29073
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against recombinant Human GAD1.

Specifications

Antigen GAD1
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against recombinant Human GAD1.
Dilution Immunocytochemistry(1:1000-1:2000) Immunohistochemistry(1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% sodium azide)
Gene Accession No. Q99259
Gene Alias FLJ45882/GAD/SCP
Gene Symbols GAD1
Host Species Chicken
Immunogen A KLH-conjugated synthetic peptide corresponding to a region near the C-terminus of GAD1 gene product, and was 100% conserved between the human (Q99259, NCBI), mouse (P48318, NCBI) and rat (NP_058703, NCBI) gene products.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2571/14415/24379
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C and avoid from light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.