Learn More
Goat IgG Horseradish Peroxidase-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | IgG |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Gene Symbols | IGHG1 |
| Host Species | Chicken |
| Immunogen | Goat IgG |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.