Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRM1, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130572 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-572 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-572 Supplier Abnova Corporation Supplier No. PAB29069
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against recombinant Human GRM1.

Specifications

Antigen GRM1
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against recombinant Human GRM1.
Dilution Immunocytochemistry(1:1000-1:2000) Immunohistochemistry(1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% sodium azide)
Gene Accession No. NP_000829.2
Gene Alias GPRC1A/GRM1A/MGLUR1/MGLUR1A/mGlu1
Gene Symbols GRM1
Host Species Chicken
Immunogen Three different KLH-conjugated synthetic peptides corresponding to different regions of GRM1 gene product, shared between the human (NP_000829.2, NCBI) and mouse (NP_058672.1, NCBI) sequences.
Purification Method Antigen affinity purification
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2911/14816
Target Species Human, Mouse
Content And Storage Store at 4°C and avoid from light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.