Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arabidopsis thaliana SEUSS, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130611 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-611 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-611 Supplier Abnova Corporation Supplier No. PAB29156

Chicken polyclonal antibody raised against Arabidopsis thaliana SEUSS.

Sequence: NQLLAEQQRNKKME

Specifications

Antigen Arabidopsis thaliana SEUSS
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against Arabidopsis thaliana SEUSS.
Dilution Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.075% sodium azide).
Gene SEU
Gene Alias F28H19.10/F28H19_10/SEUSS
Gene Symbols SEU
Host Species Chicken
Immunogen A synthetic peptide (conjugated with KLH) corresponding to internal region of Arabidopsis thaliana SEUSS emulsified in Freund's adjuvant.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 840981
Target Species Thale cress
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.