Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium botulinum Type A Neurotoxin Heavy Chain, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130609 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-609 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-609 Supplier Abnova Corporation Supplier No. PAB29131
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against Clostridium botulinum Type A Neurotoxin Heavy Chain.

Specifications

Antigen Clostridium botulinum Type A Neurotoxin Heavy Chain
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against Clostridium botulinum Type A Neurotoxin Heavy Chain.
Dilution ELISA (1:10000) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide).
Gene atx
Gene Alias bna/botA
Gene Symbols atx
Host Species Chicken
Immunogen Purified Clostridium botulinum Type A Neurotoxin Heavy Chain with Freund's adjuvant.
Purification Method Immunoaffinity Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5185061
Target Species Clostridium species
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.