Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chicken anti-IGF-IRα subunit, Polyclonal,

Catalog No. 06429IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-429-IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-429-IMI Supplier MilliporeSigma™ Supplier No. 06429I
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Specifically detects IGF-IRα subunit in Human samples, and it is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting

Insulin-like growth factor 1 receptor (EC:2.7.10.1; UniProt P08069; also known as CD221, IGF-I receptor, Insulin-like growth factor I receptor) is encoded by the IGF1R (also known as CD221, IGFIR, IGFR, JTK13) gene (Gene ID 3480) in human. The insulin-like growth factor 1 (IGF-1) receptor has tyrosine kinase that is activated by IGF-1 and IGF-2 binding. IGF-1 receptor is initially produced as a precursor protein with an aignal peptide sequence (aa 1-30). Post-translational cleavages produce one extracellular alpha (aa 31-736) chain and one transmembrane beta chain (aa 741 – 1367) linked together by a disulfide bond. Two sets of alpha-beta dimers then form the functional tetrameric receptor complex via a disulfide linkage between their alpha chain. Upon ligand binding, the alpha chains induce the tyrosine auto-phosphorylation of the beta chains, which in turn triggers cell type-specific downstream signaling cascade of events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF-IRα subunit
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Formulation Purified chicken polyclonal IgY antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P08069
Gene Symbols IGF1R
Host Species Chicken
Immunogen KLH-conjugated synthetic peptide [C-KYADGTIDIEEVTENPKT] corresponding to amino acid residues 672-689 of human IGF-IR alpha chain.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000866
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.