Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP2, Chicken, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89129480 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-480 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-480 Supplier Abnova Corporation Supplier No. PAB27965
Add to Cart
Add to Cart

Chicken polyclonal antibody against synthetic peptide of IGFBP2

Sequence: GDPECHLFYNEQQEAR

Specifications

Antigen IGFBP2
Applications Immuno-MRM (Multiple Reaction Monitoring)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Description Chicken polyclonal antibody against synthetic peptide of IGFBP2
Formulation In Phosphate-Buffered Saline with 0.02% Sodium Azide.
Gene IGFBP2
Gene Alias IBP2/IGF-BP53
Gene Symbols IGFBP2
Host Species Chicken
Immunogen A Synthetic peptide corresponding to amino acids of IGFBP2
Purification Method Antigen affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3485
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.