Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG F(ab')2, Chicken, Biotin, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against native Human IgG F(ab')2 with Biotin conjugation.
Supplier: Abnova Corporation PAB29186
Specifications
IgG F(ab')2 | |
Polyclonal | |
Chicken polyclonal antibody raised against native Human IgG F(ab')2 with Biotin conjugation. | |
In PBS, pH 7.2 (0.005% thimerosol, 50% glycerol) | |
Native purified human IgG. | |
500 μg | |
Primary | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
IgY |
ELISA, Western Blot | |
Biotin | |
ELISA Western Blot The optimal working dilution should be determined by the end user. | |
Chicken | |
Human IgG F(ab')2 affinity purification | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction