Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG F(ab')2, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89138537 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-537 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-537 Supplier Abnova Corporation Supplier No. PAB29185
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against native Human IgG F(ab')2.

Specifications

Antigen IgG F(ab')2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against native Human IgG F(ab')2.
Dilution ELISA Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.075% sodium azide)
Host Species Chicken
Immunogen Native purified human IgG.
Purification Method Human IgG F(ab')2 affinity purification
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.