Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JUN, Chicken, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89129498 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-498 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-498 Supplier Abnova Corporation Supplier No. PAB27983
Add to Cart
Add to Cart

Chicken polyclonal antibody against synthetic phosphopeptide of JUN

This gene is the putative transforming gene of avian sarcoma virus 17. It encodes a protein which is highly similar to the viral protein, and which interacts directly with specific target DNA sequences to regulate gene expression. This gene is intronless and is mapped to 1p32-p31, a chromosomal region involved in both translocations and deletions in human malignancies. [provided by RefSeq

Sequence: NSDLLT(pS)PDVGLLK

Specifications

Antigen JUN
Applications Immuno-MRM (Multiple Reaction Monitoring)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Biotin
Description Chicken polyclonal antibody against synthetic phosphopeptide of JUN
Formulation In Phosphate-Buffered Saline with 0.02% Sodium Azide.
Gene JUN
Gene Alias AP-1/AP1/c-Jun
Gene Symbols JUN
Host Species Chicken
Immunogen Synthetic phosphopeptide corresponding to amino acids of JUN
Purification Method Antigen affinity purification
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3725
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.