Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK3, Chicken, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89129495 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-495 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-495 Supplier Abnova Corporation Supplier No. PAB27980
Add to Cart
Add to Cart

Chicken polyclonal antibody against synthetic phosphopeptide of MAPK3

The protein encoded by this gene is a member of the MAP kinase family. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act in a signaling cascade that regulates various cellular processes such as proliferation, differentiation, and cell cycle progression in response to a variety of extracellular signals. This kinase is activated by upstream kinases, resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Alternatively spliced transcript variants encoding different protein isoforms have been described. [provided by RefSeq

Sequence: IADPEHDHTGFL(pT)EYVATR

Specifications

Antigen MAPK3
Applications Immuno-MRM (Multiple Reaction Monitoring)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Biotin
Description Chicken polyclonal antibody against synthetic phosphopeptide of MAPK3
Formulation In Phosphate-Buffered Saline with 0.02% Sodium Azide.
Gene MAPK3
Gene Alias ERK1/HS44KDAP/HUMKER1A/MGC20180/P44ERK1/P44MAPK/PRKM3
Gene Symbols MAPK3
Host Species Chicken
Immunogen Synthetic phosphopeptide corresponding to amino acids of MAPK3
Purification Method Antigen affinity purification
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5595
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.