Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEFH, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130584 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-584 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-584 Supplier Abnova Corporation Supplier No. PAB29081
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against recombinant Bovine NEFH.

Specifications

Antigen NEFH
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against recombinant Bovine NEFH.
Dilution Immunocytochemistry(1:2000-1:5000) Immunohistochemistry(1:2000-1:5000) Western Blot(1:5000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.2% BSA, 0.02% sodium azide)
Gene Symbols NEFH
Host Species Chicken
Immunogen Purified NEFH protein (200 kDa) isolated from bovine brains.
Purification Method IgY Fraction
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 528842/380684
Target Species Bovine, Mouse
Content And Storage Store at 4°C and avoid from light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.