Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3CA, Chicken, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89129486 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-486 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-486 Supplier Abnova Corporation Supplier No. PAB27971
Add to Cart
Add to Cart

Chicken polyclonal antibody against synthetic peptide of PIK3CA

Phosphatidylinositol 3-kinase is composed of an 85kDa regulatory subunit and a 110kDa catalytic subunit. The protein encoded by this gene represents the catalytic subunit, which uses ATP to phosphorylate PtdIns, PtdIns4P and PtdIns(4,5)P2. This gene has been found to be oncogenic and has been implicated in cervical cancers. [provided by RefSeq

Sequence: DPLSEITEQEK

Specifications

Antigen PIK3CA
Applications Immuno-MRM (Multiple Reaction Monitoring)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Description Chicken polyclonal antibody against synthetic peptide of PIK3CA
Formulation In Phosphate-Buffered Saline with 0.02% Sodium Azide.
Gene PIK3CA
Gene Alias MGC142161/MGC142163/PI3K/p110-alpha
Gene Symbols PIK3CA
Host Species Chicken
Immunogen A Synthetic peptide corresponding to amino acids of PIK3CA
Purification Method Antigen affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5290
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.