Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLP1, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130591 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-591 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-591 Supplier Abnova Corporation Supplier No. PAB29088
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against recombinant Human PLP1.

Specifications

Antigen PLP1
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against recombinant Human PLP1.
Dilution Immunocytochemistry(1:1000-1:2000) Immunohistochemistry(1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% sodium azide)
Gene Alias HLD1/MMPL/PLP/PLP/DM20/PMD/SPG2
Gene Symbols PLP1
Host Species Chicken
Immunogen A KLH-conjugated synthetic peptide corresponding to human and mouse PLP1.(NP_000524, NP_955772, P60202)
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5354/18823
Target Species Human, Mouse
Content And Storage Store at 4°C and protect from light. For long term storage, dilute the antibody with sterile glycerol and store it at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.