Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRNP, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130592 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-592 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-592 Supplier Abnova Corporation Supplier No. PAB29089
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against recombinant Human PRNP.

The protein encoded by this gene is a membrane glycosylphosphatidylinositol-anchored glycoprotein that tends to aggregate into rod-like structures. The encoded protein contains a highly unstable region of five tandem octapeptide repeats. This gene is found on chromosome 20, approximately 20kb upstream of a gene which encodes a biochemically and structurally similar protein to the one encoded by this gene. Mutations in the repeat region as well as elsewhere in this gene have been associated with Creutzfeldt-Jakob disease, fatal familial insomnia, Gerstmann-Straussler disease, Huntington disease-like 1, and kuru. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq

Specifications

Antigen PRNP
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against recombinant Human PRNP.
Dilution Immunocytochemistry(1:1000-1:5000) Immunohistochemistry(1:1000-1:5000) Western Blot(1:2500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.02% sodium azide)
Gene PRNP
Gene Alias ASCR/CD230/CJD/GSS/MGC26679/PRIP/PrP/PrP27-30/PrP33-35C/PrPc/prion
Gene Symbols PRNP
Host Species Chicken
Immunogen Recombinant fragment of human PRNP protein emulsified with Freund's adjuvant.
Purification Method Antigen affinity purification
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5621
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C and avoid from light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.