Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS6, Chicken, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89129497 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-497 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-497 Supplier Abnova Corporation Supplier No. PAB27982
Add to Cart
Add to Cart

Chicken polyclonal antibody against synthetic phosphopeptide of RPS6

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a cytoplasmic ribosomal protein that is a component of the 40S subunit. The protein belongs to the S6E family of ribosomal proteins. It is the major substrate of protein kinases in the ribosome, with subsets of five C-terminal serine residues phosphorylated by different protein kinases. Phosphorylation is induced by a wide range of stimuli, including growth factors, tumor-promoting agents, and mitogens. Dephosphorylation occurs at growth arrest. The protein may contribute to the control of cell growth and proliferation through the selective translation of particular classes of mRNA. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq

Sequence: LSSLRA(pS)TSK

Specifications

Antigen RPS6
Applications Immuno-MRM (Multiple Reaction Monitoring)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Biotin
Description Chicken polyclonal antibody against synthetic phosphopeptide of RPS6
Formulation In Phosphate-Buffered Saline with 0.02% Sodium Azide.
Gene RPS6
Gene Symbols RPS6
Host Species Chicken
Immunogen Synthetic phosphopeptide corresponding to amino acids of RPS6
Purification Method Antigen affinity purification
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6194
Target Species Human
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.