Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFTPB, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130605 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-605 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-605 Supplier Abnova Corporation Supplier No. PAB29125
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against bovine SFTPB.

Specifications

Antigen SFTPB
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against bovine SFTPB.
Dilution Western Blot (1:6000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.05% sodium azide).
Gene SFTPB
Gene Alias MGC140803
Gene Symbols SFTPB
Host Species Chicken
Immunogen Purified bovine SFTPB from Infasurf™.
Purification Method IgY fraction
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 507398
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.