Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TG with Biotin conjugation, Chicken, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89130638 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-638 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-638 Supplier Abnova Corporation Supplier No. PAB29199
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against native Human TG with Biotin conjugation.

Specifications

Antigen TG with Biotin conjugation
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Description Chicken polyclonal antibody raised against native Human TG with Biotin conjugation.
Dilution ELISA Western Blot The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide, 50% glycerol)
Gene TG
Gene Alias AITD3/TGN
Gene Symbols TG
Host Species Chicken
Immunogen Native purified human TG.
Purification Method Antigen affinity purification
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7038
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.