Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TH, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130598 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-598 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-598 Supplier Abnova Corporation Supplier No. PAB29094
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against recombinant Human TH.

Specifications

Antigen TH
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against recombinant Human TH.
Dilution Immunocytochemistry(1:1000-1:2000) Immunohistochemistry(1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% sodium azide)
Gene Alias TYH
Gene Symbols TH
Host Species Chicken
Immunogen Two different KLH-conjugated synthetic peptides corresponding to different regions of the TH gene product. These peptide sequences were shared between the mouse (P24529) and human (P07101) gene products.
Purification Method Antigen affinity purification
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7054/21823
Target Species Human, Mouse
Content And Storage Store at 4°C and avoid from light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.