Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioredoxin, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130602 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-602 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-602 Supplier Abnova Corporation Supplier No. PAB29120

Chicken polyclonal antibody raised against native Escerichia coli Thioredoxin.

Specifications

Antigen Thioredoxin
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against native Escerichia coli Thioredoxin.
Dilution ELISA (1:200-1:500 for capture; 1:10000 for primary antibody) Western Blot (1:1000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide).
Gene Alias ECK3773/JW5856/dasC/fipA/tsnC
Gene Symbols trxA
Host Species Chicken
Immunogen Native purified Escerichia coli Thioredoxin with Freund's adjuvant.
Purification Method Immunoaffinity Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 948289/1036612/3993314
Target Species E. coli
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.