Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBB3, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89130597 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
300 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-597 300 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-597 Supplier Abnova Corporation Supplier No. PAB29093
Add to Cart
Add to Cart

Chicken polyclonal antibody raised against recombinant Human TUBB3.

Specifications

Antigen TUBB3
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Chicken polyclonal antibody raised against recombinant Human TUBB3.
Dilution Immunocytochemistry(1:1000-1:2000) Immunohistochemistry(1:1000-1:2000) Western Blot(1:2000-1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% sodium azide)
Gene Alias MC1R/TUBB4/beta-4
Gene Symbols TUBB3
Host Species Chicken
Immunogen Three different KLH-conjugated synthetic peptides corresponding to different regions of the TUBB3 gene product, shared between the human (AAL28094, NCBI) and rat (AAM28438, NCBI) sequences.
Purification Method Antigen affinity purification
Quantity 300 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10381/246118
Target Species Human, Rat
Content And Storage Store at 4°C and avoid from light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.