Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WLS, Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89129501 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-501 Supplier Abnova Corporation Supplier No. PAB27986
Add to Cart
Add to Cart

Chicken polyclonal antibody against synthetic peptide of WLS

Specifications

Antigen WLS
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Chicken polyclonal antibody against synthetic peptide of WLS
Dilution Western Blot (1:2000 to 1:5000) The optimal working dilution should be determined by the end user.
Formulation In Phosphate-Buffered Saline with 0.02% Sodium Azide.
Gene GPR177
Gene Alias C1orf139/EVI/FLJ23091/MGC131760/MGC14878/MRP/WLS
Gene Symbols GPR177
Host Species Chicken
Immunogen A Synthetic peptide corresponding to C-terminus region of WLS
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79971
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage aliquot and store at -20°C. Avoid repeated freezing and thawing cycles.
Form Liquid
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.